missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Diethylaminoethyl 4-aminobenzoate hydrochloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1748530

 View more versions of this product

Catalog No. AAA1748530


Explore available promotions

Description

Description

A sodium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Diethylaminoethyl 4-aminobenzoate hydrochloride
155°C to 157°C
Odorless
MFCD00013000
UN2811
14,7757
HCBIBCJNVBAKAB-UHFFFAOYSA-N
hydrogen 2-(diethylamino)ethyl 4-aminobenzoate chloride
5795
272.78
51-05-8
195°C to 196°C (17 mmHg)
C13H21ClN2O2
250 g
Light sensitive
procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1
272.77
CHEBI:8431
99%

Chemical Identifiers

51-05-8
272.77
HCBIBCJNVBAKAB-UHFFFAOYSA-N
5795
[H+].[Cl-].CCN(CC)CCOC(=O)C1=CC=C(N)C=C1
C13H21ClN2O2
MFCD00013000
procaine hydrochloride, procaine hcl, gerovital h3, 2-diethylamino ethyl 4-aminobenzoate hydrochloride, geriocaine, atoxicocaine, chlorocaine, neotonocaine, aminocaine, anadolor
CHEBI:8431
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c

H301-H315-H317-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 200-077-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only