missing translation for 'onlineSavingsMsg'
Learn More

2-Diisopropylaminoethyl chloride hydrochloride, 98%, Thermo Scientific Chemicals

$74.67 - $1,267.49

Chemical Identifiers

CAS 4261-68-1
Molecular Formula C8H19Cl2N
Molecular Weight (g/mol) 200.15
MDL Number MFCD00012496
InChI Key IUSXYVRFJVAVOB-UHFFFAOYSA-N
Synonym 2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
PubChem CID 77942
SMILES [H+].[Cl-].CC(C)N(CCCl)C(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2266222
View Documents
Thermo Scientific Chemicals
B2266222
100 g N/A
Only null left
 
AAB2266236
View Documents
Thermo Scientific Chemicals
B2266236
500 g N/A
Only null left
 
AAB226620E
View Documents
Thermo Scientific Chemicals
B226620E
2500 g N/A
Only null left
 
Description

Description

2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Diisopropylaminoethyl chloride hydrochloride is used as an intermediate in pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

Solubility
Soluble in water.

Notes
Hygroscopic. Air sensitive. Incompatible with bases and oxidizing agents.
Specifications

Chemical Identifiers

4261-68-1
200.15
IUSXYVRFJVAVOB-UHFFFAOYSA-N
77942
C8H19Cl2N
MFCD00012496
2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
[H+].[Cl-].CC(C)N(CCCl)C(C)C

Specifications

4261-68-1
C8H19Cl2N
100 g
3551603
2-diisopropylaminoethyl chloride hydrochloride, 2-diisopropylamino ethyl chloride hydrochloride, unii-6rmr38fceq, 2-chloroethyldiisopropylamine hydrochloride, 6rmr38fceq, 2-propanamine, n-2-chloroethyl-n-1-methylethyl-, hydrochloride, 2-chloroethyldiisopropylammonium chloride, 2-chloroethyl diisopropylamine hydrochloride, n-chloroethyl diisopropylamine hydrochloride, beta-chloroethyl diisopropylamine hydrochloride
IUSXYVRFJVAVOB-UHFFFAOYSA-N
N-(2-chloroethyl)-N-propan-2-ylpropan-2-amine;hydrochloride
77942
98%
131°C to 134°C
MFCD00012496
UN2811
Hygroscopic
Soluble in water.
[H+].[Cl-].CC(C)N(CCCl)C(C)C
200.15
200.15
2-Diisopropylaminoethyl chloride hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H318-H315-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c

H301+H311-H315-H319-H330-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 224-238-1

RTECSNumber : TX1521900

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only