missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Dimethylamino)ethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™

Supplier:  TCI America A1235500G

 View more versions of this product

Catalog No. A1235500G


Explore available promotions

Description

Description

Specifications

Specifications

2-(Dimethylamino)ethyl Acrylate (stabilized with MEHQ)
Yellow
C7H13NO2
500 g
2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1
CN(C)CCOC(=O)C=C
143.19
143.19
Liquid
2439-35-2
161°C
MFCD00038233
3302
DPBJAVGHACCNRL-UHFFFAOYSA-N
2-(dimethylamino)ethyl prop-2-enoate
17111
≥98.0% (GC)

Chemical Identifiers

2439-35-2
143.19
DPBJAVGHACCNRL-UHFFFAOYSA-N
17111
CN(C)CCOC(=O)C=C
C7H13NO2
MFCD00038233
2-dimethylamino ethyl acrylate, dimethylaminoethyl acrylate, n,n-dimethylaminoethyl acrylate, adame, 2-propenoic acid, 2-dimethylamino ethyl ester, acrylic acid, 2-dimethylamino ethyl ester, 2-dimethylamino, unii-5yes121ftl, 2-n,n-dimethylamino ethyl acrylate, ageflex fa-1
2-(dimethylamino)ethyl prop-2-enoate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2583

RTECSNumber : AS8578000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions