Learn More
2-Dimethylaminoethyl acetate, 98%, Thermo Scientific Chemicals
$195.28 - $195.28
Chemical Identifiers
CAS | 1421-89-2 |
---|---|
Molecular Formula | C6H13NO2 |
Molecular Weight (g/mol) | 131.18 |
MDL Number | MFCD00041912 |
InChI Key | GOLSFPMYASLXJC-UHFFFAOYSA-N |
Synonym | 2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate |
PubChem CID | 15013 |
IUPAC Name | 2-(dimethylamino)ethyl acetate |
SMILES | CN(C)CCOC(C)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC408060250
|
Thermo Scientific Chemicals
408060250 |
25 g |
Each for $195.28
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1421-89-2 | |
131.18 | |
GOLSFPMYASLXJC-UHFFFAOYSA-N | |
15013 | |
CN(C)CCOC(C)=O |
C6H13NO2 | |
MFCD00041912 | |
2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate | |
2-(dimethylamino)ethyl acetate |
Specifications
1421-89-2 | |
100.0 | |
0.9200g/mL | |
47°C | |
98% | |
C6H13NO2 | |
CH3COOCH2CH2N(CH3)2 | |
25 g | |
2-dimethylamino ethyl acetate, dimethylaminoethyl acetate, dimethylaminoethanol acetate, 2-dimethylaminoethanol acetate, acetic acid, 2-dimethylamino ethyl ester, acetic acid 2-dimethylaminoethyl ester, 2-dimethylamino ethanol acetic ester, acmc-1c2gl, 2-dimethylaminoethyl=acetate, 2-dimethylamino ethylacetate | |
CN(C)CCOC(C)=O | |
131.18 | |
131.17 | |
Liquid |
97.5 | |
Yellow | |
152°C to 154°C | |
Authentic | |
Glass bottle | |
1.4155 to 1.4175 | |
MFCD00041912 | |
0.92 | |
GOLSFPMYASLXJC-UHFFFAOYSA-N | |
2-(dimethylamino)ethyl acetate | |
15013 | |
98% | |
2-Dimethylaminoethyl acetate, 98% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if swallowed.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 215-827-4
RTECSNumber : AH2100000
TSCA : TSCA
RUO – Research Use Only