Learn More
2-Dimethylaminoethyl chloride hydrochloride, 99%, Thermo Scientific Chemicals
$50.95 - $153.10
Chemical Identifiers
| CAS | 4584-46-7 |
|---|---|
| Molecular Formula | C4H10ClN·HCl |
| Molecular Weight (g/mol) | 144.05 |
| MDL Number | MFCD00012516 |
| InChI Key | LQLJZSJKRYTKTP-UHFFFAOYSA-N |
| Synonym | 2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride |
| PubChem CID | 11636816 |
| ChEBI | CHEBI:78153 |
| IUPAC Name | 2-chloro-N,N-dimethylethanamine;hydrochloride |
| SMILES | CN(C)CCCl.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC115821000
|
Thermo Scientific Chemicals
115821000 |
100 g | Plastic bottle |
Each for $50.95
|
|
||||
|
AC115825000
|
Thermo Scientific Chemicals
115825000 |
500 g | Plastic bottle |
Each for $153.10
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
| 4584-46-7 | |
| 144.05 | |
| LQLJZSJKRYTKTP-UHFFFAOYSA-N | |
| 11636816 | |
| 2-chloro-N,N-dimethylethanamine;hydrochloride |
| C4H10ClN·HCl | |
| MFCD00012516 | |
| 2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride | |
| CHEBI:78153 | |
| CN(C)CCCl.Cl |
Specifications
| 4584-46-7,107-99-3 | |
| 100.0 | |
| Brown-Yellow to White | |
| 99% | |
| C4H10ClN·HCl | |
| MFCD00012516 | |
| 04, 133 | |
| Solubility in water: 2000g/L (20°C). Other solubilities: soluble in alcohol (20°C) | |
| CN(C)CCCl.Cl | |
| 144.05 | |
| CHEBI:78153 | |
| 99% | |
| 2-Dimethylaminoethyl chloride hydrochloride, 99% |
| 98.5 | |
| 203.0°C to 210.0°C | |
| Authentic | |
| Plastic bottle | |
| (CH3)2NCH2CH2Cl·HCl | |
| 100 g | |
| 2-dimethylaminoethyl chloride hydrochloride, 2-chloro-n,n-dimethylethanamine hydrochloride, 2-dimethylamino ethyl chloride hydrochloride, 2-chloroethyl dimethylamine hydrochloride, 2-chloro-n,n-dimethylethylamine hydrochloride, dimethylaminoethyl chloride hydrochloride, 2-chloroethyldimethylamine hydrochloride, 1-chloro-2-dimethylaminoethane hydrochloride, 2-dimethylamino ethylchloride hydrochloride, n-2-chloroethyl dimethylamine hydrochloride | |
| LQLJZSJKRYTKTP-UHFFFAOYSA-N | |
| 2-chloro-N,N-dimethylethanamine;hydrochloride | |
| 11636816 | |
| 144.05 | |
| Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a
GHS Signal Word: Warning
EINECSNumber : 224-970-1
RTECSNumber : KQ9020000
TSCA : TSCA
RUO – Research Use Only