missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Diphenylphosphino-1-naphthoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2236103

 View more versions of this product

Catalog No. AAB2236103

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Diphenylphosphino-1-naphthoic acid
190°C to 192°C
MFCD02683090
2-diphenylphosphino-1-naphthoic acid, 2-diphenylphosphanyl naphthalene-1-carboxylic acid, acmc-20anx1, 2-diphenylphosphino-1-naphthalenecarboxylic acid, 2-diphenylphosphino-naphthalen-1-carboxylic acid, 1-naphthalenecarboxylicacid, 2-diphenylphosphino
OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
356.36
356.36
178176-80-2
C23H17O2P
1 g
WJCBDBBZIGUHOB-UHFFFAOYSA-N
2-diphenylphosphanylnaphthalene-1-carboxylic acid
10861198
97%

Chemical Identifiers

178176-80-2
356.36
WJCBDBBZIGUHOB-UHFFFAOYSA-N
10861198
OC(=O)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1
C23H17O2P
MFCD02683090
2-diphenylphosphino-1-naphthoic acid, 2-diphenylphosphanyl naphthalene-1-carboxylic acid, acmc-20anx1, 2-diphenylphosphino-1-naphthalenecarboxylic acid, 2-diphenylphosphino-naphthalen-1-carboxylic acid, 1-naphthalenecarboxylicacid, 2-diphenylphosphino
2-diphenylphosphanylnaphthalene-1-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only