missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Diphenylphosphino)biphenyl, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6164006

 View more versions of this product

Catalog No. AAH6164006


Explore available promotions

Description

Description

2-(Diphenylphosphino)biphenyl is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Diphenylphosphino)biphenyl is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

2-(Diphenylphosphino)biphenyl
C24H19P
5 g
Slightly soluble in water.
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1
338.39
338.38
Solid
13885-09-1
MFCD11559063
2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl
FNCQSSIMHQVKGF-UHFFFAOYSA-N
diphenyl-(2-phenylphenyl)phosphane
12976978
98%

Chemical Identifiers

13885-09-1
338.39
FNCQSSIMHQVKGF-UHFFFAOYSA-N
12976978
C24H19P
MFCD11559063
2-diphenylphosphino biphenyl, 2-diphenylphosphino-biphenyl, 1,1'-biphenyl-2-yldiphenylphosphane, biphenyl-2-yl-diphenyl-phosphane, phosphine, 1,1'-biphenyl yldiphenyl, diphenyl 2-phenylphenyl phosphane, 1,1'-biphenyl-2-yldiphenyl-phosphine, ksc494o1b, 2-biphenylyl diphenylphosphine, diphenylphosphino-1,1'-biphenyl
C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only