missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethoxyethyl Acetate 98.0+%, TCI America™
Supplier: TCI America E0049500ML
Specifications
2-Ethoxyethyl Acetate | |
-62°C | |
156°C | |
MFCD00009251 | |
1172 | |
SVONRAPFKPVNKG-UHFFFAOYSA-N | |
2-ethoxyethyl acetate | |
8095 | |
≥98.0% (GC) |
111-15-9 | |
Colorless | |
C6H12O3 | |
500 mL | |
ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu | |
CCOCCOC(C)=O | |
132.16 | |
132.16 | |
Liquid |
Chemical Identifiers
111-15-9 | |
132.16 | |
SVONRAPFKPVNKG-UHFFFAOYSA-N | |
8095 | |
CCOCCOC(C)=O |
C6H12O3 | |
MFCD00009251 | |
ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu | |
2-ethoxyethyl acetate |
Safety and Handling
EINECSNumber : (2)-0740
RTECSNumber : KK8225000
TSCA : Yes