Learn More
2-Ethyl-1-butene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1155314
Description
2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethyl-1-butene is used as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Soluble in alcohol, and methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C.
Specifications
2-Ethyl-1-butene | |
-132°C | |
63°C to 64°C | |
C6H12 | |
MFCD00009341 | |
UN3295 | |
2-ethyl-1-butene, 3-methylenepentane, pentane, 3-methylene, 2-ethylbut-1-ene, 1-butene, 2-ethyl, unii-o5mh5v756e, 1,1-diethylethene, acmc-1blm9, 1-butene, 2-ethyl-8ci | |
RYKZRKKEYSRDNF-UHFFFAOYSA-N | |
3-methylidenepentane | |
12970 | |
97% |
760-21-4 | |
0.689 | |
−26°C (−14°F) | |
1.396 | |
25 g | |
1697102 | |
Soluble in alcohol,and methanol. Insoluble in water. | |
CCC(=C)CC | |
84.162 | |
84.16 |
Chemical Identifiers
760-21-4 | |
84.162 | |
RYKZRKKEYSRDNF-UHFFFAOYSA-N | |
12970 | |
CCC(=C)CC |
C6H12 | |
MFCD00009341 | |
2-ethyl-1-butene, 3-methylenepentane, pentane, 3-methylene, 2-ethylbut-1-ene, 1-butene, 2-ethyl, unii-o5mh5v756e, 1,1-diethylethene, acmc-1blm9, 1-butene, 2-ethyl-8ci | |
3-methylidenepentane |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 212-078-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only