missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylamino-1-(4-methoxyphenyl)propane Hydrochloride 98.0+%, TCI America™
Supplier: TCI America E038725G
Specifications
| 2-Ethylamino-1-(4-methoxyphenyl)propane Hydrochloride | |
| 162°C | |
| C12H20ClNO | |
| 25 g | |
| KWQLOEWGKCQLPR-UHFFFAOYSA-N | |
| N-ethyl-1-(4-methoxyphenyl)propan-2-amine;hydrochloride | |
| 198672 | |
| ≥98.0% (T) |
| 93963-24-7 | |
| White-Yellow | |
| MFCD00060152 | |
| 4-(2-Ethylaminopropyl)anisole Hydrochloride | |
| CCNC(C)CC1=CC=C(C=C1)OC.Cl | |
| 229.748 | |
| 229.75 | |
| Crystalline Powder |
Chemical Identifiers
| 93963-24-7 | |
| 229.748 | |
| KWQLOEWGKCQLPR-UHFFFAOYSA-N | |
| 198672 | |
| CCNC(C)CC1=CC=C(C=C1)OC.Cl |
| C12H20ClNO | |
| MFCD00060152 | |
| 4-(2-Ethylaminopropyl)anisole Hydrochloride | |
| N-ethyl-1-(4-methoxyphenyl)propan-2-amine;hydrochloride |
Safety and Handling
TSCA : No