Learn More
2-Ethylbutyryl chloride, 99%, Thermo Scientific Chemicals
$542.71 - $542.71
Chemical Identifiers
CAS | 2736-40-5 |
---|---|
Molecular Formula | C6H11ClO |
Molecular Weight (g/mol) | 134.60 |
MDL Number | MFCD00018812 |
InChI Key | SMUKODJVMQOSAB-UHFFFAOYSA-N |
Synonym | 2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride |
PubChem CID | 75954 |
IUPAC Name | 2-ethylbutanoyl chloride |
SMILES | CCC(CC)C(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC409821000
|
Thermo Scientific Chemicals
409821000 |
100 g | Glass bottle |
Each for $542.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2736-40-5 | |
134.60 | |
SMUKODJVMQOSAB-UHFFFAOYSA-N | |
75954 | |
CCC(CC)C(Cl)=O |
C6H11ClO | |
MFCD00018812 | |
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride | |
2-ethylbutanoyl chloride |
Specifications
2736-40-5 | |
100.0 | |
0.9820g/mL | |
33°C | |
99% | |
C6H11ClO | |
(C2H5)2CHCOCl | |
100 g | |
2-ethylbutyryl chloride, 2-ethylbutyroyl chloride, butanoyl chloride, 2-ethyl, butyryl chloride, 2-ethyl, 2-ethylbutyrylchloride, diethylacetylchloride, diethylacetyl chloride, acmc-1cqal, 2-ethyl butyryl chloride, 2-ethyl-butyryl chloride | |
CCC(CC)C(Cl)=O | |
134.60 | |
134.61 | |
Liquid |
98.5 | |
Colorless | |
138°C | |
Authentic | |
Glass bottle | |
1.4230 to 1.425 | |
MFCD00018812 | |
0.982 | |
SMUKODJVMQOSAB-UHFFFAOYSA-N | |
2-ethylbutanoyl chloride | |
75954 | |
99% | |
2-Ethylbutyryl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapour.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 220-359-9
RUO – Research Use Only