missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals

$60.81 - $443.77

Chemical Identifiers

CAS 149-57-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00002675
InChI Key OBETXYAYXDNJHR-UHFFFAOYSA-N
Synonym 2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
PubChem CID 8697
IUPAC Name 2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA12644AE
View Documents
Thermo Scientific Chemicals
A12644AE
100 mL
Each for $60.81
 
AAA12644AP
View Documents
Thermo Scientific Chemicals
A12644AP
500 mL
Each for $75.62
 
AAA126440F
View Documents
Thermo Scientific Chemicals
A126440F
2500 mL
Each for $161.62
 
AAA126440D
View Documents
Thermo Scientific Chemicals
A126440D
10,000 mL
Each for $443.77
 
Description

Description

2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylhexanoic acid is used in the preparation of metal derivatives, which act as a catalyst in polymerization reactions. For example, tin 2-ethylhexanoate is used in the manufacturing of poly(lactic-co-glycolic acid). It is also used as a stabilizer for polyvinyl chlorides. It is also involved in solvent extraction and dye granulation. Further, it is used to prepare plasticizers, lubricants, detergents, flotation aids, corrosion inhibitors and alkyd resins. In addition to this, it serves as a catalyst for polyurethane foaming.

Solubility
Miscible with water.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
Specifications

Chemical Identifiers

149-57-5
144.214
OBETXYAYXDNJHR-UHFFFAOYSA-N
8697
CCCCC(CC)C(=O)O
C8H16O2
MFCD00002675
2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
2-ethylhexanoic acid

Specifications

149-57-5
0.91
226°C to 229°C
C8H16O2
MFCD00002675
1750468
Miscible with water.
CCCCC(CC)C(=O)O
144.214
144.21
2-Ethylhexanoic acid
-59°C
3
114°C (237°F)
1.425
100 mL
2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid
OBETXYAYXDNJHR-UHFFFAOYSA-N
2-ethylhexanoic acid
8697
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H302-H312
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.

P201-P202-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P310-P312-P330-P363-P501c

H302-H312-H314-H361d

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 205-743-6

RTECSNumber : MO7700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only