missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylhexyl acrylate, 99+%, stabilized, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 410110010

 View more versions of this product

Catalog No. AC410110010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethylhexyl acrylate
0.01% max. (as acrylic acid)
100.0
0.8800g/mL
86°C
99% min. (GC)
C11H20O2
CH2=CHCOOCH2CH(CH2CH3)(CH2)3CH3
1 kg
2-ethylhexyl acrylate, 2-propenoic acid, 2-ethylhexyl ester, 2-ethyl-1-hexyl acrylate, 2-ethylhexyl 2-propenoate, acrylic acid, 2-ethylhexyl ester, 2-ethylhexylacrylate, acrylic acid 2-ethylhexyl ester, 1-hexanol, 2-ethyl-, acrylate, ccris 3430, 2-ethylhexylester kyseliny akrylove
GOXQRTZXKQZDDN-UHFFFAOYSA-N
2-ethylhexyl prop-2-enoate
10 to 20ppm MEHQ
1.7 mPa.s (20°C)
184.28
Liquid
103-11-7,150-76-5
99.0
-90.0°C
215.0°C to 219.0°C
Authentic
Glass bottle
1.4340 to 1.4360
MFCD00009495
0.88
Solubility in water: insoluble
CCCCC(CC)COC(=O)C=C
184.28
7636
CHEBI:82465
99+%

Chemical Identifiers

103-11-7
184.28
GOXQRTZXKQZDDN-UHFFFAOYSA-N
7636
2-ethylhexyl prop-2-enoate
C11H20O2
MFCD00009495
2-ethylhexyl acrylate, 2-propenoic acid, 2-ethylhexyl ester, 2-ethyl-1-hexyl acrylate, 2-ethylhexyl 2-propenoate, acrylic acid, 2-ethylhexyl ester, 2-ethylhexylacrylate, acrylic acid 2-ethylhexyl ester, 1-hexanol, 2-ethyl-, acrylate, ccris 3430, 2-ethylhexylester kyseliny akrylove
CHEBI:82465
CCCCC(CC)COC(=O)C=C
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a posit

GHS Signal Word: Warning

EINECSNumber : 203-080-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only