missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylhexyl bromide, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 305335000

 View more versions of this product

Catalog No. AC305335000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Ethylhexyl bromide
18908-66-2
100.0
1.0800g/mL
69°C
94% min. (GC)
C8H17Br
CH3(CH2)3CH(C2H5)CH2Br
500 g
2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
NZWIYPLSXWYKLH-UHFFFAOYSA-N
3-(bromomethyl)heptane
86804
95%
95%
94.0
Colorless to Yellow
75.0°C to 77.0°C (16.0 mmHg)
Authentic
Glass bottle
1.4530 to 1.4550
MFCD00000220
1.08
Solubility in water: insoluble.
CCCCC(CC)CBr
193.13
193.13
Liquid

Chemical Identifiers

18908-66-2
193.13
NZWIYPLSXWYKLH-UHFFFAOYSA-N
86804
CCCCC(CC)CBr
C8H17Br
MFCD00000220
2-ethylhexyl bromide, 3-bromomethyl heptane, heptane, 3-bromomethyl, 1-bromo-2-ethylhexane, 1-bromo iso octane, 2-ethyl-bromohexane, 3-bromomethyl-heptane, 1-bromo-2 ethylhexane, 2-ethylhexylhydrobromide
3-(bromomethyl)heptane
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 242-659-9

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only