missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethylhexyl Cyanoacetate 98.0+%, TCI America™
$31.60 - $167.87
Chemical Identifiers
CAS | 13361-34-7 |
---|---|
Molecular Formula | C11H19NO2 |
Molecular Weight (g/mol) | 197.28 |
MDL Number | MFCD00019843 |
InChI Key | ZNYBQVBNSXLZNI-UHFFFAOYNA-N |
Synonym | 2-ethylhexyl cyanoacetate, acetic acid, cyano-, 2-ethylhexyl ester, cyanoacetic acid, 2-ethylhexyl ester, 2-ethylhexylcyanoacetate, cyanoacetic acid octyl ester, acetic acid, 2-cyano-, 2-ethylhexyl ester, cyanoacetic acid 2-ethylhexyl ester, ehca, thcn-nva, acmc-209bt2 |
PubChem CID | 96359 |
IUPAC Name | 2-ethylhexyl 2-cyanoacetate |
SMILES | CCCCC(CC)COC(=O)CC#N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
C138425ML
|
TCI America
C138425ML |
25 mL |
Each for $31.60
|
|
||||||
C1384500ML
|
TCI America
C1384500ML |
500 mL |
Each for $167.87
|
|
||||||
Chemical Identifiers
13361-34-7 | |
197.28 | |
ZNYBQVBNSXLZNI-UHFFFAOYNA-N | |
96359 | |
CCCCC(CC)COC(=O)CC#N |
C11H19NO2 | |
MFCD00019843 | |
2-ethylhexyl cyanoacetate, acetic acid, cyano-, 2-ethylhexyl ester, cyanoacetic acid, 2-ethylhexyl ester, 2-ethylhexylcyanoacetate, cyanoacetic acid octyl ester, acetic acid, 2-cyano-, 2-ethylhexyl ester, cyanoacetic acid 2-ethylhexyl ester, ehca, thcn-nva, acmc-209bt2 | |
2-ethylhexyl 2-cyanoacetate |
Specifications
13361-34-7 | |
C11H19NO2 | |
25 mL | |
ZNYBQVBNSXLZNI-UHFFFAOYNA-N | |
2-ethylhexyl 2-cyanoacetate | |
96359 | |
≥98.0% (GC) | |
2-Ethylhexyl Cyanoacetate |
Yellow | |
MFCD00019843 | |
2-ethylhexyl cyanoacetate, acetic acid, cyano-, 2-ethylhexyl ester, cyanoacetic acid, 2-ethylhexyl ester, 2-ethylhexylcyanoacetate, cyanoacetic acid octyl ester, acetic acid, 2-cyano-, 2-ethylhexyl ester, cyanoacetic acid 2-ethylhexyl ester, ehca, thcn-nva, acmc-209bt2 | |
CCCCC(CC)COC(=O)CC#N | |
197.28 | |
197.28 | |
Liquid |
Safety and Handling
TSCA : Yes