missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl methacrylate, 99%, stabilized

CAS: 688-84-6 | C12H22O2 | 198.31 g/mol

$178.67 - $178.67

Chemical Identifiers

CAS 688-84-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00009494
InChI Key WDQMWEYDKDCEHT-UHFFFAOYSA-N
Synonym 2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
PubChem CID 12713
IUPAC Name 2-ethylhexyl 2-methylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(=C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371170010
SDS View Documents
Thermo Scientific Chemicals
371170010
1 L Glass bottle
Each for $178.67
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

688-84-6
198.31
WDQMWEYDKDCEHT-UHFFFAOYSA-N
12713
CCCCC(CC)COC(=O)C(=C)C
C12H22O2
MFCD00009494
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
2-ethylhexyl 2-methylprop-2-enoate

Specifications

688-84-6
100.0
0.8840g/mL
92°C
98.5% min. (GC)
C12H22O2
H2C=C(CH3)CO2CH2CH(C2H5)(CH2)3CH3
1 L
2-ethylhexyl methacrylate, 2-ethyl-1-hexyl methacrylate, 2-ethylhexyl methacryate, 2-propenoic acid, 2-methyl-, 2-ethylhexyl ester, methacrylic acid 2-ethylhexyl ester, methacrylic acid, 2-ethylhexyl ester, methacrylate, 2-ethylisohexy, 2-ethylhexylmethacrylate, dsstox_cid_7293, dsstox_rid_78391
WDQMWEYDKDCEHT-UHFFFAOYSA-N
2-ethylhexyl 2-methylprop-2-enoate
40 to 60ppm MEHQ
198.31
Liquid
98.5
-50.0°C
120°C (18.0 mmHg)
Authentic
Glass bottle
1.4370 to 1.4390
MFCD00009494
0.884
Solubility in water: 0.0031g/L
CCCCC(CC)COC(=O)C(=C)C
198.31
12713
99%
2-Ethylhexyl methacrylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at r

GHS Signal Word: Warning

EINECSNumber : 211-708-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only