missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylhexyl Vinyl Ether 97.0+%, TCI America™

$250.83 - $479.92

Chemical Identifiers

CAS 103-44-6
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00053800
InChI Key DSSAWHFZNWVJEC-UHFFFAOYSA-N
Synonym Isooctyl Vinyl Ether
PubChem CID 61004
IUPAC Name 3-(ethenoxymethyl)heptane
SMILES CCCCC(CC)COC=C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I014810G
View Documents
TCI America
I014810G
10 g
Each for $250.83
 
I014825G
View Documents
TCI America
I014825G
25 g
Each for $479.92
 
Description

Description

Specifications

Chemical Identifiers

103-44-6
156.269
DSSAWHFZNWVJEC-UHFFFAOYSA-N
61004
CCCCC(CC)COC=C
C10H20O
MFCD00053800
Isooctyl Vinyl Ether
3-(ethenoxymethyl)heptane

Specifications

103-44-6
177°C
MFCD00053800
3271
DSSAWHFZNWVJEC-UHFFFAOYSA-N
3-(ethenoxymethyl)heptane
61004
≥97.0% (GC)
2-Ethylhexyl Vinyl Ether
Colorless
C10H20O
10 g
Isooctyl Vinyl Ether
CCCCC(CC)COC=C
156.269
156.27
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0372&(2)-0575

RTECSNumber : KO0175000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'