missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylnitrobenzene 99.0+%, TCI America™
$59.65 - $214.37
Chemical Identifiers
| CAS | 612-22-6 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00024294 |
| InChI Key | PXWYZLWEKCMTEZ-UHFFFAOYSA-N |
| Synonym | 2-ethylnitrobenzene, o-ethylnitrobenzene, benzene, 1-ethyl-2-nitro, ethylnitrobenzene, o-nitroethylbenzene, 2-ethyl-1-nitrobenzene, 1-ethy1-2-nitro-benzene, ccris 3099, o-ethyl nitrobenzene, benzene, ethylnitro |
| PubChem CID | 69156 |
| IUPAC Name | 1-ethyl-2-nitrobenzene |
| SMILES | CCC1=CC=CC=C1[N+](=O)[O-] |
Chemical Identifiers
| 612-22-6 | |
| 151.165 | |
| PXWYZLWEKCMTEZ-UHFFFAOYSA-N | |
| 69156 | |
| CCC1=CC=CC=C1[N+](=O)[O-] |
| C8H9NO2 | |
| MFCD00024294 | |
| 2-ethylnitrobenzene, o-ethylnitrobenzene, benzene, 1-ethyl-2-nitro, ethylnitrobenzene, o-nitroethylbenzene, 2-ethyl-1-nitrobenzene, 1-ethy1-2-nitro-benzene, ccris 3099, o-ethyl nitrobenzene, benzene, ethylnitro | |
| 1-ethyl-2-nitrobenzene |
Specifications
| 612-22-6 | |
| Yellow | |
| C8H9NO2 | |
| 25 g | |
| PXWYZLWEKCMTEZ-UHFFFAOYSA-N | |
| 1-ethyl-2-nitrobenzene | |
| 69156 | |
| ≥99.0% (GC) | |
| 2-Ethylnitrobenzene |
| -13°C | |
| 232°C | |
| MFCD00024294 | |
| 2-ethylnitrobenzene, o-ethylnitrobenzene, benzene, 1-ethyl-2-nitro, ethylnitrobenzene, o-nitroethylbenzene, 2-ethyl-1-nitrobenzene, 1-ethy1-2-nitro-benzene, ccris 3099, o-ethyl nitrobenzene, benzene, ethylnitro | |
| CCC1=CC=CC=C1[N+](=O)[O-] | |
| 151.165 | |
| 151.17 | |
| Liquid |
Safety and Handling
EINECSNumber : (9)-2453
TSCA : No