Learn More
2-Ethylphenol, 98+%, Thermo Scientific Chemicals
$67.49 - $157.81
Chemical Identifiers
CAS | 90-00-6 |
---|---|
Molecular Formula | C8H10O |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00002249 |
InChI Key | IXQGCWUGDFDQMF-UHFFFAOYSA-N |
Synonym | o-ethylphenol, phlorol, phenol, 2-ethyl, phenol, o-ethyl, ethylphenol, 1-ethyl-2-hydroxybenzene, 1-hydroxy-2-ethylbenzene, phenol, ethyl, florol, o-ethyl phenol |
PubChem CID | 6997 |
ChEBI | CHEBI:34275 |
IUPAC Name | 2-ethylphenol |
SMILES | CCC1=CC=CC=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0163709
|
Thermo Scientific Chemicals
L0163709 |
10 g |
Each for $67.49
|
|
|||||
AAL0163718
|
Thermo Scientific Chemicals
L0163718 |
50 g |
Each for $157.81
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
90-00-6 | |
122.17 | |
IXQGCWUGDFDQMF-UHFFFAOYSA-N | |
6997 | |
2-ethylphenol |
C8H10O | |
MFCD00002249 | |
o-ethylphenol, phlorol, phenol, 2-ethyl, phenol, o-ethyl, ethylphenol, 1-ethyl-2-hydroxybenzene, 1-hydroxy-2-ethylbenzene, phenol, ethyl, florol, o-ethyl phenol | |
CHEBI:34275 | |
CCC1=CC=CC=C1O |
Specifications
90-00-6 | |
1.018 | |
78°C (172°F) | |
1.536 | |
10 g | |
1099397 | |
o-ethylphenol, phlorol, phenol, 2-ethyl, phenol, o-ethyl, ethylphenol, 1-ethyl-2-hydroxybenzene, 1-hydroxy-2-ethylbenzene, phenol, ethyl, florol, o-ethyl phenol | |
CCC1=CC=CC=C1O | |
122.17 | |
CHEBI:34275 | |
≥98% |
-18°C | |
206°C | |
C8H10O | |
MFCD00002249 | |
UN3145 | |
14,3839 | |
IXQGCWUGDFDQMF-UHFFFAOYSA-N | |
2-ethylphenol | |
6997 | |
122.17 | |
2-Ethylphenol |
Safety and Handling
GHS H Statement
H314-H318-H302-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Combustible liquid.
P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, LIQUID, N.O.S.
EINECSNumber : 201-958-4
RTECSNumber : SL4025000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only