missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Fluoro-2'-deoxyadenosine, 99%, Thermo Scientific Chemicals
$244.71 - $244.71
Chemical Identifiers
CAS | 64183-27-3 |
---|---|
Molecular Formula | C10H12FN5O3 |
Molecular Weight (g/mol) | 269.24 |
MDL Number | MFCD09750859 |
InChI Key | ZGYYPTJWJBEXBC-GPGUAWMRNA-N |
Synonym | 2'-fluoro-2'-deoxyadenosine, 2'-deoxy-2'-fluoroadenosine, adenosine, 2'-deoxy-2'-fluoro, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 9-2-fluoro-2-deoxy-beta-d-ribofuranosyl-purin-6-amine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 2-fluoro-2-deoxyadenosine, 2'f-da |
PubChem CID | 100253 |
SMILES | NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6544103
|
Thermo Scientific Chemicals
J6544103 |
1 g |
Each for $244.71
|
|
|||||
Description
2'-Fluoro-2'-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2'-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2′-Fluoro-2′-deoxyadenosine is a potential prodrug for 2-fluroadenine, involves deoxygenation by reduction of the 2′-thiocar-bonylimidazolide with (Tms)3SiH as hydrogen donor.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
64183-27-3 | |
269.24 | |
ZGYYPTJWJBEXBC-GPGUAWMRNA-N | |
100253 |
C10H12FN5O3 | |
MFCD09750859 | |
2'-fluoro-2'-deoxyadenosine, 2'-deoxy-2'-fluoroadenosine, adenosine, 2'-deoxy-2'-fluoro, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 9-2-fluoro-2-deoxy-beta-d-ribofuranosyl-purin-6-amine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 2-fluoro-2-deoxyadenosine, 2'f-da | |
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1 |
Specifications
64183-27-3 | |
C10H12FN5O3 | |
1 g | |
Soluble in water. | |
NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3F)C2=NC=N1 | |
269.24 | |
269.23 | |
Powder |
White | |
MFCD09750859 | |
2'-fluoro-2'-deoxyadenosine, 2'-deoxy-2'-fluoroadenosine, adenosine, 2'-deoxy-2'-fluoro, 2r,3r,4r,5r-5-6-amino-9h-purin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 9-2-fluoro-2-deoxy-beta-d-ribofuranosyl-purin-6-amine, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl oxolan-3-ol, 2r,3r,4r,5r-5-6-aminopurin-9-yl-4-fluoro-2-hydroxymethyl tetrahydrofuran-3-ol, 2-fluoro-2-deoxyadenosine, 2'f-da | |
ZGYYPTJWJBEXBC-GPGUAWMRNA-N | |
(2R,3R,4R,5R)-5-(6-aminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol | |
100253 | |
99% | |
2'-Fluoro-2'-deoxyadenosine |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only