missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific Chemicals 2'-Fluoro-2'-deoxycytidine, 99%

Supplier:  Thermo Scientific Chemicals J6563203

Catalog No. AAJ6563203


Explore available promotions

Description

Description

2'-Fluoro-2'-deoxycytidine is a potent inhibitor of the subgenomic hepatitis C virus replicon in Huh-7 cells. 2'-Fluoro-2'-deoxycytidine has been shown to inhibit Borna Disease virus replication and spread.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Fluoro-2′-deoxycytidine is a potent inhibitor of the subgenomic hepatitis C virus replicon in Huh-7 cells. 2′-Fluoro-2′-deoxycytidine has been shown to inhibit Borna Disease virus replication and spread.
Specifications

Specifications

2'-Fluoro-2'-deoxycytidine
White
MFCD00057445
2'-deoxy-2'-fluorocytidine, 2'-fluoro-2'-deoxycytidine, 4-amino-1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl pyrimidin-2-one, cytidine, 2'-deoxy-2'-fluoro, unii-lcy080jpy9, 2-deoxy-2-fluorocytidine, lcy080jpy9, 2'-fc, 2'-fluoro-d-cytidine
NVZFZMCNALTPBY-XVFCMESISA-N
4-amino-1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
101507
99%
10212-20-1
C9H12FN3O4
1 g
Soluble in DMSO,Methanol.
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)F
245.21
245.21
Powder

Chemical Identifiers

10212-20-1
245.21
NVZFZMCNALTPBY-XVFCMESISA-N
101507
C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)F
C9H12FN3O4
MFCD00057445
2'-deoxy-2'-fluorocytidine, 2'-fluoro-2'-deoxycytidine, 4-amino-1-2r,3r,4r,5r-3-fluoro-4-hydroxy-5-hydroxymethyl oxolan-2-yl pyrimidin-2-one, cytidine, 2'-deoxy-2'-fluoro, unii-lcy080jpy9, 2-deoxy-2-fluorocytidine, lcy080jpy9, 2'-fc, 2'-fluoro-d-cytidine
4-amino-1-[(2R,3R,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only