missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-3-methylaniline 98.0+%, TCI America™
Supplier: TCI America F05545G
Specifications
2-Fluoro-3-methylaniline | |
Red | |
C7H8FN | |
5 g | |
WFZUBZAEFXETBF-UHFFFAOYSA-N | |
2-fluoro-3-methylaniline | |
22734623 | |
≥98.0% (GC,T) |
1978-33-2 | |
87°C | |
MFCD06410915 | |
3-amino-2-fluorotoluene, 2-fluoro-m-toluidine, 2-fluoro-3-methylbenzenamine, 3-amino-2-fluoro-toluene, 2-fluoro-3-methyl-aniline, 2-fluoro-3-methylphenylamine, 2-fluoro-3-aminotoluene, benzenamine, 2-fluoro-3-methyl, 2-fluoro-3-methyl-phenylamine, pubchem1536 | |
CC1=C(F)C(N)=CC=C1 | |
125.15 | |
125.15 | |
Liquid |
Chemical Identifiers
1978-33-2 | |
125.15 | |
WFZUBZAEFXETBF-UHFFFAOYSA-N | |
22734623 | |
CC1=C(F)C(N)=CC=C1 |
C7H8FN | |
MFCD06410915 | |
3-amino-2-fluorotoluene, 2-fluoro-m-toluidine, 2-fluoro-3-methylbenzenamine, 3-amino-2-fluoro-toluene, 2-fluoro-3-methyl-aniline, 2-fluoro-3-methylphenylamine, 2-fluoro-3-aminotoluene, benzenamine, 2-fluoro-3-methyl, 2-fluoro-3-methyl-phenylamine, pubchem1536 | |
2-fluoro-3-methylaniline |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator