missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-3-nitrotoluene, 98%, Thermo Scientific Chemicals

$169.10 - $536.89

Chemical Identifiers

CAS 437-86-5
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD03412242
InChI Key NBCNUIXYBLFJMI-UHFFFAOYSA-N
Synonym 2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
PubChem CID 13470957
IUPAC Name 2-fluoro-1-methyl-3-nitrobenzene
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6317314
View Documents
Thermo Scientific Chemicals
H6317314
25 g
Each for $169.10
 
AAH6317322
View Documents
Thermo Scientific Chemicals
H6317322
100 g
Each for $536.89
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

437-86-5
155.128
NBCNUIXYBLFJMI-UHFFFAOYSA-N
13470957
CC1=C(C(=CC=C1)[N+](=O)[O-])F
C7H6FNO2
MFCD03412242
2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
2-fluoro-1-methyl-3-nitrobenzene

Specifications

437-86-5
Colorless to Yellow
111°C (12 mmHg)
C7H6FNO2
MFCD03412242
UN2810
2-fluoro-3-nitrotoluene, 2-fluoro-3-methylnitrobenzene, benzene, 2-fluoro-1-methyl-3-nitro, pubchem1591, 2-fluor-3-nitrotoluol, acmc-1agoj, ksc235o7n, 2-fluoro-3-methyl-nitrobenzene, 2-fluoro-1-methyl-3-nitro-benzene, 1-methyl-2-fluoro-3-nitro-benzene
CC1=C(C(=CC=C1)[N+](=O)[O-])F
155.128
155.13
Liquid
19°C (decomposition)
1.276 g/mL
98°C (208°F)
1.529
25 g
2442082
NBCNUIXYBLFJMI-UHFFFAOYSA-N
2-fluoro-1-methyl-3-nitrobenzene
13470957
98%
2-Fluoro-3-nitrotoluene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only