missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-hydroxypyridine, 97%

CAS: 22282-69-5 | C5H4FNO | 113.091 g/mol

$171.23 - $469.58

Chemical Identifiers

CAS 22282-69-5
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD04114159
InChI Key BRLSDLHMGRDAPF-UHFFFAOYSA-N
Synonym 2-fluoropyridin-4-ol, 2-fluoro-4-hydroxypyridine, 2-fluoro-pyridin-4-ol, 4-pyridinol, 2-fluoro, 2-fluoro-4-pyridinol, 4 1h-pyridinone,2-fluoro, 2-fluoranyl-1h-pyridin-4-one, 4 1h-pyridinone, 2-fluoro, 4 1h-pyridinone, 2-fluoro-9ci
PubChem CID 2762834
IUPAC Name 2-fluoro-1H-pyridin-4-one
SMILES C1=CNC(=CC1=O)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH31697MD
View Documents
Thermo Scientific Chemicals
H31697MD
250 mg
Each for $171.23
Only null left
 
AAH3169703
View Documents
Thermo Scientific Chemicals
H3169703
1 g
Each for $469.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

22282-69-5
113.091
BRLSDLHMGRDAPF-UHFFFAOYSA-N
2762834
C1=CNC(=CC1=O)F
C5H4FNO
MFCD04114159
2-fluoropyridin-4-ol, 2-fluoro-4-hydroxypyridine, 2-fluoro-pyridin-4-ol, 4-pyridinol, 2-fluoro, 2-fluoro-4-pyridinol, 4 1h-pyridinone,2-fluoro, 2-fluoranyl-1h-pyridin-4-one, 4 1h-pyridinone, 2-fluoro, 4 1h-pyridinone, 2-fluoro-9ci
2-fluoro-1H-pyridin-4-one

Specifications

22282-69-5
C5H4FNO
250 mg
BRLSDLHMGRDAPF-UHFFFAOYSA-N
2-fluoro-1H-pyridin-4-one
2762834
97%
146°C to 149°C
MFCD04114159
2-fluoropyridin-4-ol, 2-fluoro-4-hydroxypyridine, 2-fluoro-pyridin-4-ol, 4-pyridinol, 2-fluoro, 2-fluoro-4-pyridinol, 4 1h-pyridinone,2-fluoro, 2-fluoranyl-1h-pyridin-4-one, 4 1h-pyridinone, 2-fluoro, 4 1h-pyridinone, 2-fluoro-9ci
C1=CNC(=CC1=O)F
113.091
113.09
2-Fluoro-4-hydroxypyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only