missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-4-methoxybenzoic Acid 98.0+%, TCI America™
$101.21 - $401.48
Chemical Identifiers
CAS | 394-42-3 |
---|---|
Molecular Formula | C8H7FO3 |
Molecular Weight (g/mol) | 170.139 |
MDL Number | MFCD00509373 |
InChI Key | UPWMPIKNUXTWFP-UHFFFAOYSA-N |
PubChem CID | 2774542 |
IUPAC Name | 2-fluoro-4-methoxybenzoic acid |
SMILES | COC1=CC(=C(C=C1)C(=O)O)F |
Chemical Identifiers
394-42-3 | |
170.139 | |
UPWMPIKNUXTWFP-UHFFFAOYSA-N | |
2-fluoro-4-methoxybenzoic acid |
C8H7FO3 | |
MFCD00509373 | |
2774542 | |
COC1=CC(=C(C=C1)C(=O)O)F |
Specifications
394-42-3 | |
C8H7FO3 | |
5 g | |
UPWMPIKNUXTWFP-UHFFFAOYSA-N | |
2-fluoro-4-methoxybenzoic acid | |
2774542 | |
≥98.0% (T) | |
2-Fluoro-4-methoxybenzoic Acid |
193°C | |
MFCD00509373 | |
2-fluoro-p-anisic acid, 2-fluoro-4-methoxy-benzoic acid, 2-fluoro-4-methoxybenzoicacid, benzoic acid, 2-fluoro-4-methoxy, acmc-1ajdw, 4-carboxy-3-fluoroanisole, ksc494q6n, akos brn-0650, attercop-chm at143177, benzoic acid,2-fluoro-4-methoxy | |
COC1=CC(=C(C=C1)C(=O)O)F | |
170.139 | |
170.14 | |
Crystalline Powder |
Safety and Handling
TSCA : No