missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-methoxybenzonitrile, 97%, Thermo Scientific™

$796.22 - $796.22

Chemical Identifiers

CAS 94610-82-9
Molecular Formula C8H6FNO
Molecular Weight (g/mol) 151.14
MDL Number MFCD00509374
InChI Key HWKUZTFIZATJPM-UHFFFAOYSA-N
Synonym 3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
PubChem CID 639878
IUPAC Name 2-fluoro-4-methoxybenzonitrile
SMILES COC1=CC(F)=C(C=C1)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3253106
View Documents
Thermo Scientific Chemicals
H3253106
5 g
Each for $796.22
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Fluoro-4-methoxybenzonitrile is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

94610-82-9
151.14
HWKUZTFIZATJPM-UHFFFAOYSA-N
639878
COC1=CC(F)=C(C=C1)C#N
C8H6FNO
MFCD00509374
3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
2-fluoro-4-methoxybenzonitrile

Specifications

94610-82-9
C8H6FNO
5 g
3-fluoro-4-cyanoanisole, 2-fluoro-4-methoxy-benzonitrile, benzonitrile, 2-fluoro-4-methoxy, 3-fluoro-4-cyano anisole, 4-cyano-3-fluoroanisole, buttpark 81\01-98, 2-fluoro-4-methoxybenzenecarbonitrile, pubchem4783, 2-fluoro-4-methoxybe
COC1=CC(F)=C(C=C1)C#N
151.14
151.14
2-Fluoro-4-methoxybenzonitrile
62°C to 65°C
MFCD00509374
UN3439
HWKUZTFIZATJPM-UHFFFAOYSA-N
2-fluoro-4-methoxybenzonitrile
639878
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'