Learn More
2-Fluoro-4-methylaniline, 99%, Thermo Scientific Chemicals
$197.23 - $666.56
Chemical Identifiers
CAS | 452-80-2 |
---|---|
Molecular Formula | C7H8FN |
Molecular Weight (g/mol) | 125.15 |
MDL Number | MFCD00040975 |
InChI Key | ZQEXBVHABAJPHJ-UHFFFAOYSA-N |
Synonym | 2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci |
PubChem CID | 67984 |
IUPAC Name | 2-fluoro-4-methylaniline |
SMILES | CC1=CC=C(N)C(F)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1659106
|
Thermo Scientific Chemicals
A1659106 |
5 g |
Each for $197.23
|
|
||||||
AAA1659114
|
Thermo Scientific Chemicals
A1659114 |
25 g |
Each for $666.56
|
|
||||||
Description
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.
Solubility
Not miscible in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
Chemical Identifiers
452-80-2 | |
125.15 | |
ZQEXBVHABAJPHJ-UHFFFAOYSA-N | |
67984 | |
CC1=CC=C(N)C(F)=C1 |
C7H8FN | |
MFCD00040975 | |
2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci | |
2-fluoro-4-methylaniline |
Specifications
452-80-2 | |
70°C to 71°C (7 mmHg) | |
C7H8FN | |
MFCD00040975 | |
UN2810 | |
2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci | |
ZQEXBVHABAJPHJ-UHFFFAOYSA-N | |
2-fluoro-4-methylaniline | |
67984 | |
99% |
1.108 | |
79°C (174°F) | |
1.533 | |
5 g | |
2637578 | |
Not miscible in water. | |
CC1=CC=C(N)C(F)=C1 | |
125.15 | |
125.15 | |
2-Fluoro-4-methylaniline |
Safety and Handling
GHS H Statement
H311-H373-H227-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
RTECSNumber : XU6650000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only