missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-4-nitrophenol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1313414

 View more versions of this product

Catalog No. AAA1313414


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluoro-4-nitrophenol
117°C to 121°C
MFCD00051970
1944995
ORPHLVJBJOCHBR-UHFFFAOYSA-N
157.1
157.1
403-19-0
C6H4FNO3
25 g
2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t
C1=CC(=C(C=C1[N+](=O)[O-])F)O
9825
≥98%

Chemical Identifiers

403-19-0
157.1
ORPHLVJBJOCHBR-UHFFFAOYSA-N
9825
C6H4FNO3
MFCD00051970
2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t
C1=CC(=C(C=C1[N+](=O)[O-])F)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only