missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxypyridine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3299403

 View more versions of this product

Catalog No. AAH3299403


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Fluoro-6-methoxypyridine
C6H6FNO
1 g
pyridine, 2-fluoro-6-methoxy, acmc-1bnhh, 2-fluoro-6-methoxy-pyridine, pyridine,2-fluoro-6-methoxy, 2-methoxy-6-fluoropyridine
COC1=NC(=CC=C1)F
127.118
127.12
116241-61-3
MFCD09998161
UN1993
PKZOFECOOLMHMG-UHFFFAOYSA-N
2-fluoro-6-methoxypyridine
14296367
97%

Chemical Identifiers

116241-61-3
127.118
PKZOFECOOLMHMG-UHFFFAOYSA-N
2-fluoro-6-methoxypyridine
C6H6FNO
MFCD09998161
14296367
COC1=NC(=CC=C1)F
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only