missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-m-xylene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1042414

 View more versions of this product

Catalog No. AAA1042414


Explore available promotions

Description

Description

It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the spectroscopic observation of jet-cooled 2-fluoro-m-xylyl radical.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Specifications

Specifications

2-Fluoro-m-xylene
0.988
25 g
1.479
MFCD00039215
1857413
Not miscible or difficult to mix in water.
CC1=C(C(=CC=C1)C)F
124.158
124.16
443-88-9
147°C to 148°C
Aromatic
C8H9F
UN1993
2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
JTAUTNBVFDTYTI-UHFFFAOYSA-N
2-fluoro-1,3-dimethylbenzene
123065
98%

Chemical Identifiers

443-88-9
124.158
JTAUTNBVFDTYTI-UHFFFAOYSA-N
123065
CC1=C(C(=CC=C1)C)F
C8H9F
MFCD00039215
2,6-dimethylfluorobenzene, 2-fluoro-m-xylene, 1-fluoro-2,6-dimethylbenzene, 1,3-dimethyl-2-fluorobenzene, benzene, 2-fluoro-1,3-dimethyl, 2,6-dimethyl fluorobenzene, 2-fluoro-1,3-dimethyl-benzene, fr b1 f1, pubchem4407, 2-fluoro-3-methyltoluene
2-fluoro-1,3-dimethylbenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 207-144-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only