missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroaniline, 99%

CAS: 348-54-9 | C6H6FN | 111.12 g/mol

$50.32 - $400.73

Chemical Identifiers

CAS 348-54-9
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007642
InChI Key FTZQXOJYPFINKJ-UHFFFAOYSA-N
Synonym o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
PubChem CID 9584
ChEBI CHEBI:27526
IUPAC Name 2-fluoroaniline
SMILES NC1=CC=CC=C1F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1261214
View Documents
Thermo Scientific Chemicals
A1261214
25 g
Each for $50.32
Only null left
 
AAA1261222
View Documents
Thermo Scientific Chemicals
A1261222
100 g
Each for $126.05
Only null left
 
AAA1261236
View Documents
Thermo Scientific Chemicals
A1261236
500 g
Each for $400.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

348-54-9
111.12
FTZQXOJYPFINKJ-UHFFFAOYSA-N
9584
2-fluoroaniline
C6H6FN
MFCD00007642
o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
CHEBI:27526
NC1=CC=CC=C1F

Specifications

348-54-9
1.152
60°C (140°F)
C6H6FN
MFCD00007642
UN2941
o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
NC1=CC=CC=C1F
111.12
CHEBI:27526
99%
-29°C
171°C to 173°C
Recognizable
1.542
25 g
1524219
FTZQXOJYPFINKJ-UHFFFAOYSA-N
2-fluoroaniline
9584
111.12
2-Fluoroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H226-H373-H302-H312-H315-H319
Toxic if inhaled.
Flammable liquid and vapour.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302+H312+H332-H315-H318-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES

EINECSNumber : 206-478-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only