Learn More
2-Fluorobiphenyl, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1222709
Description
2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Fluorobiphenyl is used as pharmaceuticals, veterinary drugs and organic synthesis intermediates. 2-Fluorobiphenyl was used as an internal standard for analysis of environmental pollutants in sediments using pressurized liquid extraction and gas chromatography-mass spectrometry, as a sole carbon & energy source for P. pseudoalcaligenes.
Solubility
Soluble in alcohol, ether. Insoluble in water.
Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
2-Fluorobiphenyl | |
72°C to 76°C | |
248°C | |
MFCD00000317 | |
2043175 | |
Soluble in alcohol,ether. Insoluble in water. | |
FC1=CC=CC=C1C1=CC=CC=C1 | |
172.20 | |
172.2 |
321-60-8 | |
1.245 | |
C12H9F | |
10 g | |
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
KLECYOQFQXJYBC-UHFFFAOYSA-N | |
2-fluoro-1,1'-biphenyl | |
67579 | |
98% |
Chemical Identifiers
321-60-8 | |
172.20 | |
KLECYOQFQXJYBC-UHFFFAOYSA-N | |
67579 |
C12H9F | |
MFCD00000317 | |
2-fluorobiphenyl, 2-fluoro-1,1'-biphenyl, o-fluorodiphenyl, 1,1'-biphenyl, 2-fluoro, biphenyl, 2-fluoro, ortho-fluorodiphenyl, 2-fluorodiphenyl, 2-fluoro-biphenyl, ccris 1659, 2-fluoro-1-phenylbenzene | |
FC1=CC=CC=C1C1=CC=CC=C1 |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 206-290-7
RTECSNumber : DV5291000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only