missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorophenethylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 52721-69-4
Molecular Formula C8H11FN
Molecular Weight (g/mol) 140.18
MDL Number MFCD00075502
InChI Key RIKUOLJPJNVTEP-UHFFFAOYSA-O
Synonym 2-fluorophenethylamine, 2-2-fluorophenyl ethanamine, 2-2-fluorophenyl ethylamine, benzeneethanamine, 2-fluoro, 2-2-fluoro-phenyl-ethylamine, 2-2-fluorophenyl ethan-1-amine, 2-fluoro-phenethylamine, 2-fluorophenethyl amine, 2-fluorobenzeneethanamine, benzeneethanamine,2-fluoro
PubChem CID 643357
IUPAC Name 2-(2-fluorophenyl)ethan-1-aminium
SMILES [NH3+]CCC1=CC=CC=C1F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC350930050
SDS View Documents
Thermo Scientific Chemicals
350930050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

52721-69-4
140.18
RIKUOLJPJNVTEP-UHFFFAOYSA-O
643357
[NH3+]CCC1=CC=CC=C1F
C8H11FN
MFCD00075502
2-fluorophenethylamine, 2-2-fluorophenyl ethanamine, 2-2-fluorophenyl ethylamine, benzeneethanamine, 2-fluoro, 2-2-fluoro-phenyl-ethylamine, 2-2-fluorophenyl ethan-1-amine, 2-fluoro-phenethylamine, 2-fluorophenethyl amine, 2-fluorobenzeneethanamine, benzeneethanamine,2-fluoro
2-(2-fluorophenyl)ethan-1-aminium

Specifications

52721-69-4
100.0
64°C (0.6mmHg)
Authentic
Glass bottle
1.5100 to 1.512
MFCD00075502
1.066
RIKUOLJPJNVTEP-UHFFFAOYSA-O
2-(2-fluorophenyl)ethan-1-aminium
643357
97%
98.5
1.0660g/mL
77°C
96% min. (GC)
C8H11FN
FC6H4CH2CH2NH2
5g
2-fluorophenethylamine, 2-2-fluorophenyl ethanamine, 2-2-fluorophenyl ethylamine, benzeneethanamine, 2-fluoro, 2-2-fluoro-phenyl-ethylamine, 2-2-fluorophenyl ethan-1-amine, 2-fluoro-phenethylamine, 2-fluorophenethyl amine, 2-fluorobenzeneethanamine, benzeneethanamine,2-fluoro
[NH3+]CCC1=CC=CC=C1F
140.18
139.17
2-Fluorophenethylamine, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only