missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluorophenylacetonitrile, 97%, Thermo Scientific Chemicals

$46.84 - $378.56

Chemical Identifiers

CAS 326-62-5
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00001897
InChI Key DAVJMKMVLKOQQC-UHFFFAOYSA-N
Synonym 2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
PubChem CID 67592
IUPAC Name 2-(2-fluorophenyl)acetonitrile
SMILES FC1=CC=CC=C1CC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1161206
View Documents
Thermo Scientific Chemicals
A1161206
5 g N/A
 
AAA1161214
View Documents
Thermo Scientific Chemicals
A1161214
25 g N/A
 
AAA1161222
View Documents
Thermo Scientific Chemicals
A1161222
100 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

326-62-5
135.14
DAVJMKMVLKOQQC-UHFFFAOYSA-N
67592
FC1=CC=CC=C1CC#N
C8H6FN
MFCD00001897
2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
2-(2-fluorophenyl)acetonitrile

Specifications

326-62-5
235°C
C8H6FN
MFCD00001897
UN3276
2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
FC1=CC=CC=C1CC#N
135.14
135.14
2-Fluorophenylacetonitrile
1.061
>100°C (212°F)
1.501
5 g
1862361
DAVJMKMVLKOQQC-UHFFFAOYSA-N
2-(2-fluorophenyl)acetonitrile
67592
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c

H226-H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 206-313-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only