missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Fmoc-amino)isobutyric acid, 98%, Thermo Scientific Chemicals
$872.92 - $872.92
Chemical Identifiers
CAS | 94744-50-0 |
---|---|
Molecular Formula | C19H19NO4 |
Molecular Weight (g/mol) | 325.36 |
MDL Number | MFCD00151913 |
InChI Key | HOZZVEPRYYCBTO-UHFFFAOYSA-N |
Synonym | fmoc-aib-oh, fmoc-alpha-methylalanine, fmoc-alpha-me-ala-oh, 2-fmoc-amino isobutyric acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-2-methylpropanoic acid, alanine, n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methylpropanoic acid, n-fmoc-2-aminoisobutyric acid, 2-9h-fluoren-9-ylmethoxy carbonylamino isobutyric acid |
PubChem CID | 2756096 |
SMILES | CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6221114
|
Thermo Scientific Chemicals
H6221114 |
25 g |
Each for $872.92
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
94744-50-0 | |
325.36 | |
HOZZVEPRYYCBTO-UHFFFAOYSA-N | |
2756096 |
C19H19NO4 | |
MFCD00151913 | |
fmoc-aib-oh, fmoc-alpha-methylalanine, fmoc-alpha-me-ala-oh, 2-fmoc-amino isobutyric acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-2-methylpropanoic acid, alanine, n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methylpropanoic acid, n-fmoc-2-aminoisobutyric acid, 2-9h-fluoren-9-ylmethoxy carbonylamino isobutyric acid | |
CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
Specifications
94744-50-0 | |
C19H19NO4 | |
25 g | |
Moisture sensitive | |
HOZZVEPRYYCBTO-UHFFFAOYSA-N | |
2-(9H-fluoren-9-ylmethoxycarbonylamino)-2-methylpropanoic acid | |
2756096 | |
98% |
182°C to 188°C | |
MFCD00151913 | |
5604328 | |
fmoc-aib-oh, fmoc-alpha-methylalanine, fmoc-alpha-me-ala-oh, 2-fmoc-amino isobutyric acid, 2-9h-fluoren-9-yl methoxy carbonyl amino-2-methylpropanoic acid, alanine, n-9h-fluoren-9-ylmethoxy carbonyl-2-methyl, 2-9h-fluoren-9-ylmethoxy carbonyl amino-2-methylpropanoic acid, n-fmoc-2-aminoisobutyric acid, 2-9h-fluoren-9-ylmethoxy carbonylamino isobutyric acid | |
CC(C)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
325.36 | |
325.36 | |
2-(Fmoc-amino)isobutyric acid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only