missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Formylbenzeneboronic acid, 97%, Thermo Scientific Chemicals

$57.52 - $268.45

Chemical Identifiers

CAS 40138-16-7
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151822
InChI Key DGUWACLYDSWXRZ-UHFFFAOYSA-N
Synonym 2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde, 2-dihydroxyboryl benzaldehyde, boronic acid, 2-formylphenyl, 2-benzaldehyde boronic acid, 2-formylphenyl-boronic acid
PubChem CID 292189
IUPAC Name (2-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2543403
View Documents
Thermo Scientific Chemicals
B2543403
1 g
Each for $57.52
 
AAB2543406
View Documents
Thermo Scientific Chemicals
B2543406
5 g
Each for $268.45
 
Description

Description

2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

40138-16-7
149.94
DGUWACLYDSWXRZ-UHFFFAOYSA-N
292189
OB(O)C1=CC=CC=C1C=O
C7H7BO3
MFCD00151822
2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde, 2-dihydroxyboryl benzaldehyde, boronic acid, 2-formylphenyl, 2-benzaldehyde boronic acid, 2-formylphenyl-boronic acid
(2-formylphenyl)boronic acid

Specifications

40138-16-7
C7H7BO3
1 g
Air Sensitive
Soluble in methanol.
OB(O)C1=CC=CC=C1C=O
149.94
149.94
2-Formylbenzeneboronic acid
∼115°C to 120°C
MFCD00151822
3030776
2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde, 2-dihydroxyboryl benzaldehyde, boronic acid, 2-formylphenyl, 2-benzaldehyde boronic acid, 2-formylphenyl-boronic acid
DGUWACLYDSWXRZ-UHFFFAOYSA-N
(2-formylphenyl)boronic acid
292189
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only