missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Furoyl chloride, 98+%, Thermo Scientific Chemicals

$100.02 - $100.02

Chemical Identifiers

CAS 527-69-5
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.53
MDL Number MFCD00003228
InChI Key OFTKFKYVSBNYEC-UHFFFAOYSA-N
Synonym 2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
PubChem CID 68242
IUPAC Name furan-2-carbonyl chloride
SMILES C1=COC(=C1)C(=O)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC165380250
SDS View Documents
Thermo Scientific Chemicals
165380250
25 mL Glass bottle
Each for $100.02
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

527-69-5
130.53
OFTKFKYVSBNYEC-UHFFFAOYSA-N
68242
C1=COC(=C1)C(=O)Cl
C5H3ClO2
MFCD00003228
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
furan-2-carbonyl chloride

Specifications

527-69-5
100.0
Yellow to Brown
173°C to 174°C
Authentic
Glass bottle
1.5300 to 1.532
25 mL
1.32
2-furoyl chloride, furoyl chloride, 2-furancarbonyl chloride, furancarbonyl chloride, 2-furoylchloride, unii-2us0dxi75s, 2us0dxi75s, 2-furancarboxylic acid chloride, furan-2-carboxylic acid chloride, furanoyl chloride
OFTKFKYVSBNYEC-UHFFFAOYSA-N
furan-2-carbonyl chloride
68242
98+%
2-Furoyl chloride, 98+%
98.0
-2°C
1.3200g/mL
85°C
98% min. (GC)
C5H3ClO2
MFCD00003228
18, 276
15, 4339
Solubility in water: decomposes. Other solubilities: soluble in ether,toluene,methylene chloride,,dichloroethane,chlorobenzene,ethyl acetate,thf,and chloroform,decomposes in alcohol
C1=COC(=C1)C(=O)Cl
130.53
130.53
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 208-422-9

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only