missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hexyne, 98+%, Thermo Scientific Chemicals

$396.33 - $396.33

Chemical Identifiers

CAS 764-35-2
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00009470
InChI Key MELUCTCJOARQQG-UHFFFAOYSA-N
Synonym 2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3
PubChem CID 33629
IUPAC Name hex-2-yne
SMILES CCCC#CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2240518
View Documents
Thermo Scientific Chemicals
B2240518
50 g
Each for $396.33
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

764-35-2
82.146
MELUCTCJOARQQG-UHFFFAOYSA-N
33629
CCCC#CC
C6H10
MFCD00009470
2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3
hex-2-yne

Specifications

764-35-2
0.736
−10°C (14°F)
C6H10
MFCD00009470
UN3295
2-hexyne, methyl propyl acetylene, methylpropylacetylene, acmc-209p3f, 1-methyl-2-propylacetylene, c3h7c.$.cch3
CCCC#CC
82.146
82.15
2-Hexyne
-90°C
84°C to 86°C
Characteristic
1.414
50 g
1731542
MELUCTCJOARQQG-UHFFFAOYSA-N
hex-2-yne
33629
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H304-H315-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 212-117-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only