missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-1-naphthoic Acid 98.0+%, TCI America™
$65.24 - $361.62
Chemical Identifiers
| CAS | 2283-08-1 |
|---|---|
| Molecular Formula | C11H8O3 |
| Molecular Weight (g/mol) | 188.18 |
| MDL Number | MFCD00004009 |
| InChI Key | UPHOPMSGKZNELG-UHFFFAOYSA-N |
| Synonym | 2-hydroxy-1-naphthoic acid, 1-naphthalenecarboxylic acid, 2-hydroxy, hydroxynaphthoic acid, 1-naphthoic acid, 2-hydroxy, unii-e3h4qz94uy, e3h4qz94uy, 1-naphthalenecarboxylic acid, hydroxy, 2-hydroxynaphthalenecarboxylic acid, 2-hydroxy-1-naphthoicacid, 2-hydroxynaphtoic acid |
| PubChem CID | 16790 |
| IUPAC Name | 2-hydroxynaphthalene-1-carboxylic acid |
| SMILES | C1=CC=C2C(=C1)C=CC(=C2C(=O)O)O |
Chemical Identifiers
| 2283-08-1 | |
| 188.18 | |
| UPHOPMSGKZNELG-UHFFFAOYSA-N | |
| 16790 | |
| C1=CC=C2C(=C1)C=CC(=C2C(=O)O)O |
| C11H8O3 | |
| MFCD00004009 | |
| 2-hydroxy-1-naphthoic acid, 1-naphthalenecarboxylic acid, 2-hydroxy, hydroxynaphthoic acid, 1-naphthoic acid, 2-hydroxy, unii-e3h4qz94uy, e3h4qz94uy, 1-naphthalenecarboxylic acid, hydroxy, 2-hydroxynaphthalenecarboxylic acid, 2-hydroxy-1-naphthoicacid, 2-hydroxynaphtoic acid | |
| 2-hydroxynaphthalene-1-carboxylic acid |
Specifications
| 2283-08-1 | |
| C11H8O3 | |
| 25 g | |
| UPHOPMSGKZNELG-UHFFFAOYSA-N | |
| 2-hydroxynaphthalene-1-carboxylic acid | |
| 16790 | |
| ≥98.0% (HPLC,T) | |
| 2-Hydroxy-1-naphthoic Acid |
| 167°C | |
| MFCD00004009 | |
| 2-hydroxy-1-naphthoic acid, 1-naphthalenecarboxylic acid, 2-hydroxy, hydroxynaphthoic acid, 1-naphthoic acid, 2-hydroxy, unii-e3h4qz94uy, e3h4qz94uy, 1-naphthalenecarboxylic acid, hydroxy, 2-hydroxynaphthalenecarboxylic acid, 2-hydroxy-1-naphthoicacid, 2-hydroxynaphtoic acid | |
| C1=CC=C2C(=C1)C=CC(=C2C(=O)O)O | |
| 188.18 | |
| 188.18 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0398
TSCA : Yes