missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxy-2-azaadamantane 98.0+%, TCI America™
$184.54 - $2018.78
Chemical Identifiers
| CAS | 1155843-79-0 |
|---|---|
| Molecular Formula | C9H15NO |
| Molecular Weight (g/mol) | 153.225 |
| InChI Key | SFXINVPZIZMFHM-UHFFFAOYSA-N |
| Synonym | AZADOL(regR) |
| PubChem CID | 51037224 |
| SMILES | C1C2CC3CC1CC(C2)N3O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
H1404200MG
|
TCI America
H1404200MG |
200 mg |
Each for $184.54
|
|
|||||
|
H14041G
|
TCI America
H14041G |
1 g |
Each for $553.62
|
|
|||||
|
H14045G
|
TCI America
H14045G |
5 g |
Each for $2,018.78
|
|
|||||
Chemical Identifiers
| 1155843-79-0 | |
| 153.225 | |
| AZADOL(regR) | |
| C1C2CC3CC1CC(C2)N3O |
| C9H15NO | |
| SFXINVPZIZMFHM-UHFFFAOYSA-N | |
| 51037224 |
Specifications
| 1155843-79-0 | |
| 200 mg | |
| SFXINVPZIZMFHM-UHFFFAOYSA-N | |
| 153.225 | |
| 153.23 | |
| Crystalline Powder |
| C9H15NO | |
| AZADOL(regR) | |
| C1C2CC3CC1CC(C2)N3O | |
| 51037224 | |
| ≥98.0% (GC,T) | |
| 2-Hydroxy-2-azaadamantane |
Safety and Handling
TSCA : No