missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxy-3-methylbenzaldehyde, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 398540050

 View more versions of this product

Catalog No. AC398540050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Hydroxy-3-methylbenzaldehyde
97.5
Orange to Yellow
203°C to 205°C (730.0 mmHg)
Authentic
Glass bottle
CH3C6H3(OH)CHO
5 g
3-methylsalicylaldehyde, 2,3-cresotaldehyde, 2-hydroxy-3-methyl-benzaldehyde, 3-methylsalicylic aldehyde, 3-methyl-2-hydroxybenzaldehyde, benzaldehyde, hydroxy-3-methyl, benzaldehyde, 2-hydroxy-3-methyl, zlchem 747, pubchem14264, 2-formyl-6-methylphenol
CC1=CC=CC(=C1O)C=O
136.15
CHEBI:20110
98%
824-42-0
100.0
1.1340g/mL
87°C
97.5% min. (GC)
C8H8O2
MFCD00040798
1.134
IPPQNXSAJZOTJZ-UHFFFAOYSA-N
2-hydroxy-3-methylbenzaldehyde
522777
136.15
Liquid

Chemical Identifiers

824-42-0
136.15
IPPQNXSAJZOTJZ-UHFFFAOYSA-N
522777
2-hydroxy-3-methylbenzaldehyde
C8H8O2
MFCD00040798
3-methylsalicylaldehyde, 2,3-cresotaldehyde, 2-hydroxy-3-methyl-benzaldehyde, 3-methylsalicylic aldehyde, 3-methyl-2-hydroxybenzaldehyde, benzaldehyde, hydroxy-3-methyl, benzaldehyde, 2-hydroxy-3-methyl, zlchem 747, pubchem14264, 2-formyl-6-methylphenol
CHEBI:20110
CC1=CC=CC(=C1O)C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only