missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methoxyaniline Hydrochloride 98.0+%, TCI America™

$173.37 - $548.03

Chemical Identifiers

CAS 39547-15-4
Molecular Formula C7H10ClNO2
Molecular Weight (g/mol) 175.61
MDL Number MFCD00052002
InChI Key ZXJIQOBYRBTOLH-UHFFFAOYSA-N
Synonym 2-Amino-5-methoxyphenol Hydrochloride, 2-Hydroxy-p-anisidine Hydrochloride
PubChem CID 12210184
IUPAC Name 2-amino-5-methoxyphenol hydrochloride
SMILES Cl.COC1=CC(O)=C(N)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A12591G
View Documents
TCI America
A12591G
1 g
Each for $173.37
Only null left
 
A12595G
View Documents
TCI America
A12595G
5 g
Each for $548.03
Only null left
 
Description

Description

Specifications

Chemical Identifiers

39547-15-4
175.61
ZXJIQOBYRBTOLH-UHFFFAOYSA-N
12210184
Cl.COC1=CC(O)=C(N)C=C1
C7H10ClNO2
MFCD00052002
2-Amino-5-methoxyphenol Hydrochloride, 2-Hydroxy-p-anisidine Hydrochloride
2-amino-5-methoxyphenol hydrochloride

Specifications

39547-15-4
C7H10ClNO2
1 g
ZXJIQOBYRBTOLH-UHFFFAOYSA-N
2-amino-5-methoxyphenol hydrochloride
12210184
≥98.0% (N)
2-Hydroxy-4-methoxyaniline Hydrochloride
213°C
MFCD00052002
2-Amino-5-methoxyphenol Hydrochloride, 2-Hydroxy-p-anisidine Hydrochloride
Cl.COC1=CC(O)=C(N)C=C1
175.61
175.61
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'