missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-5-nitrobenzaldehyde, 98+%

CAS: 97-51-8 | C7H5NO4 | 167.12 g/mol

$83.50 - $277.25

Chemical Identifiers

CAS 97-51-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007337
InChI Key IHFRMUGEILMHNU-UHFFFAOYSA-N
Synonym 5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
PubChem CID 66808
IUPAC Name 2-hydroxy-5-nitrobenzaldehyde
SMILES OC1=CC=C(C=C1C=O)[N+]([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC416180050
SDS View Documents
Thermo Scientific Chemicals
416180050
5 g
Each for $83.50
Only null left
 
AC416180250
SDS View Documents
Thermo Scientific Chemicals
416180250
25 g
Each for $277.25
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97-51-8
167.12
IHFRMUGEILMHNU-UHFFFAOYSA-N
66808
OC1=CC=C(C=C1C=O)[N+]([O-])=O
C7H5NO4
MFCD00007337
5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
2-hydroxy-5-nitrobenzaldehyde

Specifications

97-51-8
100.0
Brown to Yellow
98% min. (GC)
C7H5NO4
5 g
Solubility in water: slightly soluble (20°C)
OC1=CC=C(C=C1C=O)[N+]([O-])=O
167.12
167.12
Crystals or Powder
98.0
125.0°C to 129.0°C
Authentic
Glass bottle
MFCD00007337
5-nitrosalicylaldehyde, benzaldehyde, 2-hydroxy-5-nitro, salicylaldehyde, 5-nitro, 5-nitrosaliclaldehyde, 2-hydroxy-5-nitro-benzaldehyde, 5-nitro salicylaldehdye, timtec-bb sbb003885, labotest-bb lt00920994, salicylaldehyde, 5-nitro-6ci,7ci,8ci, pubchem2052
IHFRMUGEILMHNU-UHFFFAOYSA-N
2-hydroxy-5-nitrobenzaldehyde
66808
98+%
2-Hydroxy-5-nitrobenzaldehyde, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-587-0

RTECSNumber : CU6675000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only