Learn More
2-Hydroxy-6-methoxybenzaldehyde, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1314506
Description
2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxy-6-methoxybenzaldehyde is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications
2-Hydroxy-6-methoxybenzaldehyde | |
68°C to 74°C | |
MFCD00151830 | |
1101524 | |
6-methoxysalicylaldehyde, 2-hydroxy-6-methoxy-benzaldehyde, 2-hydroxy-6-methoxybenzal dehyde, 2-hydroxy-6-methoxy benzaldehyde, benzaldehyde, 2-hydroxy-6-methoxy, pubchem8210, 2-methoxy-6-hydroxybenzaldehyde, benzaldehyde,2-hydroxy-6-methoxy, 2-hydroxy-6-methyloxy benzaldehyde | |
DZJPDDVDKXHRLF-UHFFFAOYSA-N | |
2-hydroxy-6-methoxybenzaldehyde | |
69692 | |
≥98% |
700-44-7 | |
C8H8O3 | |
5 g | |
Air Sensitive | |
Slightly soluble in water. | |
COC1=CC=CC(=C1C=O)O | |
152.149 | |
152.15 |
Chemical Identifiers
700-44-7 | |
152.149 | |
DZJPDDVDKXHRLF-UHFFFAOYSA-N | |
2-hydroxy-6-methoxybenzaldehyde |
C8H8O3 | |
MFCD00151830 | |
69692 | |
COC1=CC=CC(=C1C=O)O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 211-844-6
TSCA : No
RUO – Research Use Only