missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyethyl acrylate, 97%, stab. with 200-300ppm 4-methoxyphenol, Thermo Scientific Chemicals

$105.74 - $294.01

Chemical Identifiers

CAS 818-61-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00002865
InChI Key OMIGHNLMNHATMP-UHFFFAOYSA-N
Synonym 2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
PubChem CID 13165
IUPAC Name 2-hydroxyethyl prop-2-enoate
SMILES C=CC(=O)OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2481530
View Documents
Thermo Scientific Chemicals
B2481530
250 g
Each for $105.74
 
AAB248150B
View Documents
Thermo Scientific Chemicals
B248150B
1000 g
Each for $294.01
 
Description

Description

As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As a reactive monomer2-Hydroxyethyl acrylate is used as a crosslinking agent for resins, plastics and rubber modifiers. Further, it is used in the synthesis of amphilic block copolymers by nitroxide mediated living radical polymerization. In addition to this, it is used to prepare tuned poly(hydroxyethyl acrylate) by atom transfer radical polymerization.

Solubility
Miscible with water.

Notes
Light sensitive. Incompatible with acids, bases, peroxides, metals, copper and oxidizing agents.
Specifications

Chemical Identifiers

818-61-1
116.116
OMIGHNLMNHATMP-UHFFFAOYSA-N
13165
C=CC(=O)OCCO
C5H8O3
MFCD00002865
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
2-hydroxyethyl prop-2-enoate

Specifications

818-61-1
1.106
101°C (214°F)
C5H8O3
CH2=CHCO2CH2CH2OH
250 g
Light sensitive
Miscible with water.
C=CC(=O)OCCO
116.116
116.12
2-Hydroxyethyl acrylate, stabilized with 200 to 300ppm 4-methoxyphenol
<-60°C
192°C
Pungent
1.45
MFCD00002865
UN2927
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl prop-2-enoate
13165
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H314-H317
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

P260-P262-P264b-P270-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P330-P333+P313-P362-P363-P501c

H302-H310-H314-H317

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.

EINECSNumber : 212-454-9

RTECSNumber : AT1750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only