missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Hydroxyethyl acrylate, 97%, stabilized, Thermo Scientific Chemicals

$103.37 - $280.12

Chemical Identifiers

CAS 818-61-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002865
InChI Key OMIGHNLMNHATMP-UHFFFAOYSA-N
Synonym 2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
PubChem CID 13165
IUPAC Name 2-hydroxyethyl prop-2-enoate
SMILES C=CC(=O)OCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC334082500
SDS View Documents
Thermo Scientific Chemicals
334082500
250 mL Glass bottle
Each for $103.37
 
AC334080010
SDS View Documents
Thermo Scientific Chemicals
334080010
1 L Glass bottle
Each for $280.12
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

818-61-1
116.12
OMIGHNLMNHATMP-UHFFFAOYSA-N
13165
C=CC(=O)OCCO
C5H8O3
MFCD00002865
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
2-hydroxyethyl prop-2-enoate

Specifications

818-61-1,150-76-5
96.0
-60°C
1.1060g/mL
99°C
96% min. (GC)
C5H8O3
MFCD00002865
1.106
Solubility in water: soluble in water
C=CC(=O)OCCO
116.12
13165
116.12
Oily Liquid
4mg KOH/g max.
100.0
Yellow to Brown
210°C to 215°C
Authentic
Glass bottle
1.4490 to 1.451
250 mL
2-hydroxyethyl acrylate, hydroxyethyl acrylate, 2-propenoic acid, 2-hydroxyethyl ester, ethylene glycol monoacrylate, bisomer 2hea, acrylic acid 2-hydroxyethyl ester, 2-acryloyloxy ethanol, ethylene glycol, acrylate, acrylic acid, 2-hydroxyethyl ester, 2-hydroxyethylacrylate
OMIGHNLMNHATMP-UHFFFAOYSA-N
2-hydroxyethyl prop-2-enoate
200 to 450ppm MEHQ
9 mPa.s (20°C)
97%
2-Hydroxyethyl acrylate, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Toxic in contact with skin.
Very toxic to aquatic life.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-454-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only