missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals

$59.10 - $727.08

Chemical Identifiers

CAS 109-84-2
Molecular Formula C2H8N2O
Molecular Weight (g/mol) 76.099
MDL Number MFCD00007623
InChI Key GBHCABUWWQUMAJ-UHFFFAOYSA-N
Synonym 2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
PubChem CID 8017
ChEBI CHEBI:50098
IUPAC Name 2-hydrazinylethanol
SMILES C(CO)NN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4379309
View Documents
Thermo Scientific Chemicals
04379309
10 g
Each for $59.10
 
AA4379318
View Documents
Thermo Scientific Chemicals
04379318
50 g
Each for $172.22
 
AA4379330
View Documents
Thermo Scientific Chemicals
04379330
250 g
Each for $727.08
 
Description

Description

(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-84-2
76.099
GBHCABUWWQUMAJ-UHFFFAOYSA-N
8017
2-hydrazinylethanol
C2H8N2O
MFCD00007623
2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
CHEBI:50098
C(CO)NN

Specifications

109-84-2
1.121 g/mL
73°C (163°F)
1.493
10 g
2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
GBHCABUWWQUMAJ-UHFFFAOYSA-N
2-hydrazinylethanol
8017
76.1
(2-Hydroxyethyl)hydrazine
-70°C
218°C to 220°C
C2H8N2O
MFCD00007623
UN2810
Soluble in water,alcohols and glycols.
C(CO)NN
76.099
CHEBI:50098
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 203-711-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only