missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Hydroxyflavanone, 98%, Thermo Scientific Chemicals

$173.14 - $597.23

Chemical Identifiers

CAS 35244-11-2
Molecular Formula C15H12O3
MDL Number MFCD00017702
Synonym 2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, 2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2503903
View Documents
Thermo Scientific Chemicals
B2503903
1 g
Each for $173.14
 
AAB2503906
View Documents
Thermo Scientific Chemicals
B2503906
5 g
Each for $597.23
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35244-11-2
C15H12O3
MFCD00017702
2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, 2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one

Specifications

35244-11-2
MFCD00017702
2r-2-2-hydroxyphenyl-2,3-dihydrochromen-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, 2r-2-2-hydroxyphenyl-3,4-dihydro-2h-1-benzopyran-4-one, 2r-2-2-hydroxyphenyl-2,3-dihydro-1-benzopyran-4-one
98%
C15H12O3
1 g
240.25
2'-Hydroxyflavanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only