missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Hydroxymethyl)-2-nitro-1,3-propanediol 99.0+%, TCI America™
$65.17 - $321.92
Chemical Identifiers
CAS | 126-11-4 |
---|---|
Molecular Formula | C4H9NO5 |
Molecular Weight (g/mol) | 151.12 |
MDL Number | MFCD00007395 |
InChI Key | OLQJQHSAWMFDJE-UHFFFAOYSA-N |
Synonym | tris hydroxymethyl nitromethane, 2-hydroxymethyl-2-nitropropane-1,3-diol, 2-hydroxymethyl-2-nitro-1,3-propanediol, cimcool wafers, tris nitro, trimethylolnitromethane, isobutylglycerol, nitro, nitroisobutylglycerol, 1,3-propanediol, 2-hydroxymethyl-2-nitro, methane, trimethylolnitro |
PubChem CID | 31337 |
IUPAC Name | 2-(hydroxymethyl)-2-nitropropane-1,3-diol |
SMILES | OCC(CO)(CO)[N+]([O-])=O |
Chemical Identifiers
126-11-4 | |
151.12 | |
OLQJQHSAWMFDJE-UHFFFAOYSA-N | |
31337 | |
OCC(CO)(CO)[N+]([O-])=O |
C4H9NO5 | |
MFCD00007395 | |
tris hydroxymethyl nitromethane, 2-hydroxymethyl-2-nitropropane-1,3-diol, 2-hydroxymethyl-2-nitro-1,3-propanediol, cimcool wafers, tris nitro, trimethylolnitromethane, isobutylglycerol, nitro, nitroisobutylglycerol, 1,3-propanediol, 2-hydroxymethyl-2-nitro, methane, trimethylolnitro | |
2-(hydroxymethyl)-2-nitropropane-1,3-diol |
Specifications
126-11-4 | |
White-Yellow | |
MFCD00007395 | |
tris hydroxymethyl nitromethane, 2-hydroxymethyl-2-nitropropane-1,3-diol, 2-hydroxymethyl-2-nitro-1,3-propanediol, cimcool wafers, tris nitro, trimethylolnitromethane, isobutylglycerol, nitro, nitroisobutylglycerol, 1,3-propanediol, 2-hydroxymethyl-2-nitro, methane, trimethylolnitro | |
OCC(CO)(CO)[N+]([O-])=O | |
151.12 | |
151.12 | |
Crystalline Powder |
214°C | |
C4H9NO5 | |
25 g | |
OLQJQHSAWMFDJE-UHFFFAOYSA-N | |
2-(hydroxymethyl)-2-nitropropane-1,3-diol | |
31337 | |
≥99.0% (GC) | |
2-(Hydroxymethyl)-2-nitro-1,3-propanediol |
Safety and Handling
EINECSNumber : (9)-0900
RTECSNumber : TY7350000
TSCA : Yes