Learn More
2-Hydroxyphenethyl alcohol, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 182490010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2-Hydroxyphenethyl alcohol | |
| 97.5 | |
| 1.1590g/mL | |
| Authentic | |
| Glass bottle | |
| 1.5560 to 1.5600 (20°C, 589nm) | |
| MFCD00002890 | |
| 06,906 | |
| 2-hydroxyphenethyl alcohol, 2-2-hydroxyethyl phenol, 2-hydroxyphenylethanol, o-2-hydroxyethyl phenol, benzeneethanol, 2-hydroxy, 2-2-hydroxyphenyl ethanol, o-hydroxyphenethyl alcohol, 2-o-hydroxyphenyl ethanol, 2-hydroxy-benzeneethanol, beta-o-hydroxyphenyl ethanol | |
| OCCC1=CC=CC=C1O | |
| 138.17 | |
| CHEBI:64803 | |
| 98% |
| 7768-28-7 | |
| 100.0 | |
| 168°C to 169°C (2.0mmHg) | |
| 97.5% min. (GC) | |
| C8H10O2 | |
| HOC6H4CH2CH2OH | |
| 1g | |
| 1.159 | |
| ABFCOJLLBHXNOU-UHFFFAOYSA-N | |
| 2-(2-hydroxyethyl)phenol | |
| 82200 | |
| 138.17 |
Chemical Identifiers
| 7768-28-7 | |
| 138.17 | |
| ABFCOJLLBHXNOU-UHFFFAOYSA-N | |
| 82200 | |
| 2-(2-hydroxyethyl)phenol |
| C8H10O2 | |
| MFCD00002890 | |
| 2-hydroxyphenethyl alcohol, 2-2-hydroxyethyl phenol, 2-hydroxyphenylethanol, o-2-hydroxyethyl phenol, benzeneethanol, 2-hydroxy, 2-2-hydroxyphenyl ethanol, o-hydroxyphenethyl alcohol, 2-o-hydroxyphenyl ethanol, 2-hydroxy-benzeneethanol, beta-o-hydroxyphenyl ethanol | |
| CHEBI:64803 | |
| OCCC1=CC=CC=C1O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
EINECSNumber : 231-863-3
RUO â Research Use Only